Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025165.2 | NP_079441.1 | 7 | 1138 | ||
GenBank mRNA | AF276512.1 | - | 7 | 735 | ||
AK026290.1 | - | 7 | 820 | |||
AK057528.1 | - | 5 | 823 | |||
AK126384.1 | - | 7 | 1377 | |||
BC006548.2 | - | 7 | 789 | 2821972 | ||
BC019293.1 | - | 7 | 789 | 2821972 | ||
CR457358.1 | - | 7 | 735 |