Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025208.4 | NP_079484.1 | 2 | 638 | ||
NM_033135.3 | NP_149126.1 | 2 | 620 | |||
GenBank mRNA | AB033832.1 | - | 2 | 422 | ||
AF113216.1 | - | 2 | 372 | |||
AF335584.1 | - | 2 | 447 | |||
AF336376.1 | - | 2 | 441 | |||
AK292801.1 | - | 2 | 446 | |||
AY027517.1 | - | 2 | 358 | |||
AY027518.1 | - | 2 | 340 | |||
AY359116.1 | - | 2 | 279 | |||
BC030645.2 | - | 2 | 700 | 4824526 | ||
BG722948.1 | - | 2 | 510 |