Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136538.1 | NP_001130010.1 | 4 | 628 | ||
NM_025247.5 | NP_079523.3 | 4 | 628 | |||
GenBank mRNA | AK092356.1 | - | 4 | 613 | ||
AK310923.1 | - | 4 | 613 | |||
AL832043.1 | - | 4 | 606 | |||
AY323912.1 | - | 4 | 584 | |||
BC015056.1 | - | 4 | 600 | |||
BC126358.1 | - | 4 | 542 | 8992074 |