Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK075395.1 | - | 5, 6 | 528 | ||
AK312905.1 | - | 5, 6 | 499 | |||
AY358640.1 | - | 5, 6 | 508 | |||
BC036460.2 | - | 5, 6 | 571 | 4814497 | ||
BC056874.1 | - | 5, 6 | 510 | 5740203 |