Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171184.1 | NP_001164655.1 | 12, 13 | 1621 | ||
NM_001939.2 | NP_001930.2 | 14, 15 | 1904 | |||
XM_017029333.1 | XP_016884822.1 | 13, 14 | 1987 | |||
XM_017029334.1 | XP_016884823.1 | 13, 14 | 1983 | |||
GenBank mRNA | AB209305.1 | - | 5, 6 | 501 | ||
AK289825.1 | - | 14, 15 | 1792 | |||
AK295843.1 | - | 12, 13 | 1490 | |||
BC111695.1 | - | 14, 15 | 1741 | 40036070 | ||
U43519.1 | - | 14, 15 | 1853 |