Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030777.3 | NP_110404.1 | 5 | 3560 | ||
XM_011529060.2 | XP_011527362.1 | 5 | 3643 | |||
XM_011529061.2 | XP_011527363.1 | 5 | 5200 | |||
XM_011529062.2 | XP_011527364.1 | 4 | 3567 | |||
XM_011529065.2 | XP_011527367.1 | 4 | 3514 | |||
XM_017028087.1 | XP_016883576.1 | 4 | 3417 | |||
GenBank mRNA | AF248053.1 | - | 5 | 3712 | ||
AF321240.1 | - | 5 | 3515 | |||
AL137188.3 | - | 4 | 3471 |