Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030780.4 | NP_110407.2 | 4 | 938 | ||
NR_102337.1 | 3 | 852 | ||||
NR_102338.1 | 5 | 1047 | ||||
XM_017013877.1 | XP_016869366.1 | 4 | 841 | |||
GenBank mRNA | AF283645.1 | - | 4 | 668 | ||
AK027531.1 | - | 4 | 707 | |||
AK027787.1 | - | 4 | 672 | |||
AK125492.1 | - | 5 | 656 | |||
AK300748.1 | - | 3 | 586 | |||
BC021893.1 | - | 4 | 646 | 4524907 | ||
DC331449.1 | - | 3 | 350 |