Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030780.4 | NP_110407.2 | 6 | 1165 | ||
NR_102337.1 | 5 | 1079 | ||||
NR_102338.1 | 7 | 1274 | ||||
XM_017013877.1 | XP_016869366.1 | 6 | 1068 | |||
GenBank mRNA | AF283645.1 | - | 6 | 895 | ||
AK027531.1 | - | 6 | 934 | |||
AK027787.1 | - | 6 | 899 | |||
AK125492.1 | - | 7 | 883 | |||
AK300748.1 | - | 5 | 813 | |||
BC021893.1 | - | 6 | 873 | 4524907 |