Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030912.2 | NP_112174.2 | 6 | 2455 | ||
GenBank mRNA | AF086326.1 | - | 1 | 86 | ||
AF220034.1 | - | 6 | 2332 | |||
AF281046.1 | - | 6 | 2341 | |||
BC021925.1 | - | 6 | 2454 | 4857393 |