Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001941.4 | NP_001932.2 | 9 | 1306 | ||
NM_024423.3 | NP_077741.2 | 9 | 1306 | |||
GenBank mRNA | AK291748.1 | - | 9 | 1232 | ||
D17427.1 | - | 9 | 1218 | |||
X83929.1 | - | 10 | 1172 |