Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146008.1 | NP_001139480.1 | 8 | 3491 | ||
NM_001146009.1 | NP_001139481.1 | 8 | 3170 | |||
NM_030958.2 | NP_112220.2 | 10 | 3546 | |||
XM_005251313.2 | XP_005251370.1 | 9 | 3381 | |||
XM_017013883.1 | XP_016869372.1 | 9 | 3481 | |||
XM_017013885.1 | XP_016869374.1 | 9 | 3189 | |||
GenBank mRNA | AF205075.1 | - | 10 | 3455 | ||
AK022760.1 | - | 8 | 3491 | |||
BC016067.2 | - | 1 | 749 | |||
BC144618.1 | - | 5 | 2626 |