Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030961.2 | NP_112223.1 | 3 | 2748 | ||
XM_011516589.2 | XP_011514891.1 | 2 | 2600 | |||
GenBank mRNA | AK075255.1 | - | 1 | 1168 | ||
AL512757.1 | - | 1 | 738 | |||
BC005847.2 | - | 3 | 2747 | 2907142 | ||
BC048194.1 | - | 3 | 2682 | 5814702 |