Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130011.1 | NP_001123483.1 | 4 | 461 | ||
NM_031451.4 | NP_113639.4 | 7 | 881 | |||
XM_005259303.3 | XP_005259360.2 | 4 | 451 | |||
GenBank mRNA | AF241268.1 | - | 7 | 880 | ||
AF353396.1 | - | 4 | 551 | |||
AY014285.1 | - | 4 | 433 | |||
AY359077.1 | - | 4 | 429 | |||
BC001861.2 | - | 5 | 628 | 3538494 |