Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031914.2 | NP_114120.2 | 1 | 370 | ||
XM_006720272.3 | XP_006720335.1 | 3 | 830 | |||
XM_011537219.1 | XP_011535521.1 | 3 | 907 | |||
XM_011537220.1 | XP_011535522.1 | 3 | 907 | |||
XM_011537221.2 | XP_011535523.1 | 3 | 907 | |||
XM_011537222.2 | XP_011535524.1 | 3 | 907 | |||
XM_017021693.1 | XP_016877182.1 | 3 | 907 | |||
XM_017021694.1 | XP_016877183.1 | 3 | 596 | |||
XM_017021695.1 | XP_016877184.1 | 4 | 741 | |||
GenBank mRNA | AB102949.1 | - | 1 | 173 | ||
AJ617628.1 | - | 1 | 371 | |||
AK302919.1 | - | 2 | 670 | |||
BC117363.1 | - | 1 | 229 | 40125914 | ||
BC143598.1 | - | 1 | 229 | 9052110 |