Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122979.2 | NP_001116451.1 | 8 | 1212 | ||
NM_001170631.1 | NP_001164102.1 | 8 | 2097 | |||
NM_032029.4 | NP_114418.2 | 7 | 1295 | |||
XM_011510051.2 | XP_011508353.1 | 9 | 2046 | |||
XM_011510052.2 | XP_011508354.1 | 8 | 1807 | |||
XM_017002442.1 | XP_016857931.1 | 9 | 2042 | |||
GenBank mRNA | AF354295.1 | - | 7 | 1492 | ||
AK298394.1 | - | 8 | 2073 | |||
AY063125.1 | - | 6 | 1480 | |||
AY063126.1 | - | 2 | 1181 | |||
BC109263.1 | - | 7 | 819 | |||
BC109264.2 | - | 7 | 819 |