Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168319.1 | NP_001161791.1 | 4 | 826 | ||
NM_001955.4 | NP_001946.3 | 4 | 829 | |||
XM_011514330.2 | XP_011512632.1 | 5 | 585 | |||
XM_011514331.2 | XP_011512633.1 | 6 | 746 | |||
XM_011514332.2 | XP_011512634.1 | 5 | 583 | |||
XM_017010331.1 | XP_016865820.1 | 5 | 769 | |||
GenBank mRNA | AK226096.1 | - | 4 | 744 | ||
AK291838.1 | - | 4 | 766 | |||
AK313914.1 | - | 4 | 761 | |||
BC009720.1 | - | 4 | 698 | 3900704 | ||
BX439515.2 | - | 4 | 789 | |||
CR541903.1 | - | 4 | 495 | |||
S56805.1 | - | 4 | 831 | |||
Y00749.1 | - | 4 | 747 |