Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287498.1 | NP_001274427.1 | 14 | 2383 | ||
NM_032295.3 | NP_115671.1 | 12 | 2129 | |||
NM_207113.2 | NP_996996.1 | 15 | 2431 | |||
XM_011516626.2 | XP_011514928.1 | 16 | 2776 | |||
XM_011516627.2 | XP_011514929.1 | 15 | 2727 | |||
XM_017012712.1 | XP_016868201.1 | 17 | 2904 | |||
XM_017012713.1 | XP_016868202.1 | 14 | 2621 | |||
XM_017012714.1 | XP_016868203.1 | 15 | 2701 | |||
XM_017012715.1 | XP_016868204.1 | 15 | 2683 | |||
XM_017012716.1 | XP_016868205.1 | 12 | 2385 | |||
XM_017012717.1 | XP_016868206.1 | 13 | 2479 | |||
GenBank mRNA | AK074823.1 | - | 15 | 2392 | ||
AK126589.1 | - | 3 | 1575 | |||
AL136583.1 | - | 12 | 2036 | |||
BC028380.1 | - | 15 | 2381 | 4811814 | ||
BC046567.1 | - | 14 | 2335 | 5246993 |