Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032303.4 | NP_115679.2 | 3, 4 | 543 | ||
NR_036651.1 | 2, 3 | 379 | ||||
XM_011519091.2 | XP_011517393.1 | 3, 4 | 543 | |||
XM_017015204.1 | XP_016870693.1 | 3, 4 | 1160 | |||
XM_017015205.1 | XP_016870694.1 | 2, 3 | 996 | |||
GenBank mRNA | AK098721.1 | - | 3, 4 | 514 | ||
AK289929.1 | - | 3, 4 | 371 | |||
AK301469.1 | - | 3, 4 | 340 | |||
AK303645.1 | - | 2, 3 | 185 | |||
AL833735.1 | - | 1, 2 | 57 | |||
AY093428.1 | - | 3, 4 | 382 | |||
BC095451.1 | - | 3, 4 | 320 | 30405073 | ||
DB052480.1 | - | 3, 4 | 547 |