Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195822.1 | NP_001182751.1 | 7 | 998 | ||
NM_032303.4 | NP_115679.2 | 9 | 1217 | |||
NR_036651.1 | 8 | 1053 | ||||
XM_017015203.1 | XP_016870692.1 | 6 | 1451 | |||
XM_017015204.1 | XP_016870693.1 | 8 | 1730 | |||
XM_017015205.1 | XP_016870694.1 | 7 | 1566 | |||
GenBank mRNA | AK090940.1 | - | 1 | 13 | ||
AK098721.1 | - | 9 | 1188 | |||
AK225946.1 | - | 6 | 750 | |||
AK289929.1 | - | 9 | 1045 | |||
AK292486.1 | - | 7 | 805 | |||
AK301469.1 | - | 7 | 811 | |||
AK303645.1 | - | 8 | 859 | |||
AK313605.1 | - | 7 | 805 | |||
AL833735.1 | - | 7 | 731 | |||
AY093428.1 | - | 9 | 1056 | |||
BC004331.1 | - | 7 | 827 | 3630835 | ||
BC095451.1 | - | 9 | 994 | 30405073 |