Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001956.4 | NP_001947.1 | 4, 5 | 533 | ||
NR_126098.1 | 4, 5 | 442 | ||||
GenBank mRNA | BC034393.1 | - | 4, 5 | 476 | 5185394 | |
BI761655.1 | - | 4, 5 | 431 | |||
M65199.1 | - | 4, 5 | 521 | |||
S63835.1 | - | 2, 3 | 146 | |||
S63837.1 | - | 3, 4 | 269 | |||
S63838.1 | - | 4, 5 | 321 | |||
X55177.1 | - | 4, 5 | 509 |