Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184743.1 | NP_001171672.1 | 7 | 1454 | ||
NM_032507.3 | NP_115896.1 | 7 | 1434 | |||
XM_006715228.2 | XP_006715291.1 | 6 | 1104 | |||
XM_011514942.2 | XP_011513244.1 | 6 | 1085 | |||
XM_011514943.2 | XP_011513245.1 | 6 | 1070 | |||
XM_011514944.2 | XP_011513246.1 | 6 | 1689 | |||
XM_017011361.1 | XP_016866850.1 | 6 | 1111 | |||
XM_017011362.1 | XP_016866851.1 | 7 | 1220 | |||
XM_017011363.1 | XP_016866852.1 | 7 | 1988 | |||
GenBank mRNA | AK223446.1 | - | 7 | 1419 | ||
AK291228.1 | - | 7 | 1399 | |||
BC032507.1 | - | 7 | 1384 | |||
BC069033.1 | - | 7 | 1280 | |||
BC128585.1 | - | 7 | 1281 | 40128523 | ||
D88259.1 | - | 6 | 1014 |