Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303512.1 | NP_001290441.1 | 8 | 3653 | ||
NM_001303513.1 | NP_001290442.1 | 9 | 3896 | |||
NM_001303514.1 | NP_001290443.1 | 6 | 3308 | |||
NM_001303515.1 | NP_001290444.1 | 8 | 3488 | |||
NM_001303516.1 | NP_001290445.1 | 8 | 3470 | |||
NM_032512.3 | NP_115901.2 | 8 | 3635 | |||
GenBank mRNA | AB103478.1 | - | 8 | 3616 | ||
AF085862.1 | - | 1 | 371 | |||
AK091444.1 | - | 8 | 3361 | |||
AK123822.1 | - | 8 | 3485 | |||
AK127016.1 | - | 8 | 3632 | |||
BC002606.2 | - | 1 | 1766 | |||
BC080584.1 | - | 1 | 1766 |