Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032583.3 | NP_115972.2 | 5 | 500 | ||
NM_033151.3 | NP_149163.2 | 4 | 786 | |||
NM_145186.2 | NP_660187.1 | 5 | 500 | |||
XM_017023795.1 | XP_016879284.1 | 5 | 3114 | |||
XM_017023796.1 | XP_016879285.1 | 6 | 587 | |||
XM_017023797.1 | XP_016879286.1 | 6 | 715 | |||
XM_017023798.1 | XP_016879287.1 | 5 | 541 | |||
XM_017023799.1 | XP_016879288.1 | 5 | 523 | |||
XM_017023800.1 | XP_016879289.1 | 5 | 514 | |||
XM_017023801.1 | XP_016879290.1 | 5 | 3114 | |||
XM_017023803.1 | XP_016879292.1 | 5 | 3114 | |||
GenBank mRNA | AF352582.1 | - | 5 | 535 | ||
AF367202.1 | - | 5 | 514 | |||
AF411579.1 | - | 5 | 514 | |||
AK301547.1 | - | 5 | 541 | |||
AY040219.1 | - | 4 | 786 |