Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172674.1 | NP_001166145.1 | 5 | 3585 | ||
NM_001172675.1 | NP_001166146.1 | 5 | 3265 | |||
NM_032584.2 | NP_115973.2 | 5 | 3262 | |||
XM_005259335.4 | XP_005259392.1 | 5 | 3584 | |||
XM_005259336.4 | XP_005259393.1 | 5 | 3374 | |||
XM_017027384.1 | XP_016882873.1 | 5 | 3604 | |||
XM_017027385.1 | XP_016882874.1 | 5 | 3288 | |||
XM_017027386.1 | XP_016882875.1 | 4 | 3210 | |||
GenBank mRNA | AL713691.1 | - | 1 | 2243 | ||
AY029765.1 | - | 5 | 3224 |