Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001962.2 | NP_001953.1 | 5 | 1490 | ||
XM_006714565.2 | XP_006714628.1 | 4 | 2878 | |||
XM_011543250.2 | XP_011541552.1 | 5 | 1303 | |||
XM_011543251.2 | XP_011541553.1 | 5 | 2816 | |||
XM_017009205.1 | XP_016864694.1 | 5 | 2579 | |||
GenBank mRNA | U26403.1 | - | 5 | 1490 |