Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032709.2 | NP_116098.2 | 5 | 423 | ||
XM_011540293.2 | XP_011538595.1 | 6 | 1845 | |||
XM_017016835.1 | XP_016872324.1 | 5 | 384 | |||
XM_017016836.1 | XP_016872325.1 | 5 | 384 | |||
XM_017016837.1 | XP_016872326.1 | 5 | 384 | |||
XM_017016838.1 | XP_016872327.1 | 5 | 2619 | |||
XM_017016839.1 | XP_016872328.1 | 3 | 476 | |||
XM_017016840.1 | XP_016872329.1 | 3 | 330 | |||
XM_017016841.1 | XP_016872330.1 | 2 | 637 | |||
GenBank mRNA | AK074429.1 | - | 5 | 384 | ||
AK075265.1 | - | 5 | 361 | |||
AK225637.1 | - | 5 | 377 | |||
AK295699.1 | - | 5 | 396 | |||
BC006131.1 | - | 5 | 386 | 3626506 | ||
DC361966.1 | - | 5 | 424 |