Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322080.1 | NP_001309009.1 | 5 | 691 | ||
NM_001322081.1 | NP_001309010.1 | 5 | 592 | |||
NM_032718.4 | NP_116107.3 | 5 | 575 | |||
NR_136173.1 | 6 | 659 | ||||
NR_136174.1 | 5 | 544 | ||||
XM_011512005.2 | XP_011510307.1 | 5 | 698 | |||
XM_011512006.2 | XP_011510308.1 | 5 | 515 | |||
XM_011512007.2 | XP_011510309.1 | 3 | 172 | |||
GenBank mRNA | AK075376.1 | - | 5 | 541 | ||
AK290589.1 | - | 5 | 529 | |||
AK296768.1 | - | 5 | 691 | |||
AK307178.1 | - | 6 | 649 | |||
BC006242.2 | - | 5 | 544 | 3952602 |