Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032782.4 | NP_116171.3 | 2 | 468 | ||
GenBank mRNA | AB924452.1 | - | 1 | 151 | ||
AF251707.1 | - | 2 | 340 | |||
AF450242.1 | - | 2 | 271 | |||
AF450243.1 | - | 2 | 271 | |||
AK027334.1 | - | 2 | 257 | |||
AK314406.1 | - | 2 | 275 | |||
AY069944.1 | - | 2 | 214 | |||
BC020843.1 | - | 2 | 323 | |||
BC063431.1 | - | 2 | 358 | 5229539 | ||
BP291914.1 | - | 2 | 468 | |||
JX049979.1 | - | 2 | 214 |