Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048164.2 | NP_001041629.1 | 8, 9 | 1444 | ||
NM_032803.5 | NP_116192.4 | 8, 9 | 1468 | |||
XM_017029912.1 | XP_016885401.1 | 7, 8 | 1959 | |||
GenBank mRNA | AF320612.1 | - | 8, 9 | 1419 | ||
AK027447.1 | - | 6, 7 | 790 | |||
AK074865.1 | - | 8, 9 | 1439 | |||
AK075014.1 | - | 8, 9 | 1444 | |||
BC033816.1 | - | 8, 9 | 1423 | 5206252 |