Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033452.2 | NP_258411.2 | 6 | 1387 | ||
XM_005257787.4 | XP_005257844.1 | 6 | 1434 | |||
XM_005257788.4 | XP_005257845.1 | 6 | 793 | |||
GenBank mRNA | AY026763.1 | - | 6 | 1388 | ||
BC006153.1 | - | 5 | 656 | |||
BC009225.2 | - | 3 | 246 | |||
BC017299.2 | - | 6 | 819 | 5017710 |