Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002211.3 | NP_002202.2 | 14 | 2218 | ||
NM_033668.2 | NP_391988.1 | 13 | 1997 | |||
NM_133376.2 | NP_596867.1 | 14 | 2133 | |||
GenBank mRNA | AK291697.1 | - | 14 | 2070 | ||
BC020057.1 | - | 14 | 2062 | 3924411 | ||
BX537407.1 | - | 14 | 2072 | |||
DA600874.1 | - | 2 | 231 |