Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMTN SIRNA Assays ›
Gene Symbol:
DMTN
Gene Name:
dematin actin binding protein
Gene Aliases:
DMT, EPB49
Chromosome Location:
Chr.8: 22048955 - 22082527 on Build GRCh38
UniGene ID:
Hs.106124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
10989
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114135.4
NP_001107607.1
9, 10
998
NM_001114136.2
NP_001107608.1
9, 10
954
NM_001114137.3
NP_001107609.1
9, 10
1128
NM_001114138.2
NP_001107610.1
9, 10
1115
NM_001114139.3
NP_001107611.1
8, 9
1012
NM_001302816.2
NP_001289745.1
9, 10
1248
NM_001302817.2
NP_001289746.1
7, 8
967
NM_001323378.1
NP_001310307.1
9, 10
1087
NM_001323380.1
NP_001310309.1
9, 10
994
NM_001323381.1
NP_001310310.1
8, 9
1103
NM_001323382.1
NP_001310311.1
9, 10
1033
NM_001323383.1
NP_001310312.1
9, 10
954
NM_001323384.1
NP_001310313.1
9, 10
1087
NM_001323385.1
NP_001310314.1
8, 9
1103
NM_001323387.1
NP_001310316.1
8, 9
1052
NM_001323388.1
NP_001310317.1
7, 8
834
NM_001323389.1
NP_001310318.1
8, 9
1042
NM_001323390.1
NP_001310319.1
8, 9
909
NM_001323391.1
NP_001310320.1
8, 9
1012
NM_001323392.1
NP_001310321.1
8, 9
1173
NM_001323393.1
NP_001310322.1
8, 9
879
NM_001323394.1
NP_001310323.1
8, 9
1040
NM_001323395.1
NP_001310324.1
8, 9
919
NM_001323396.1
NP_001310325.1
8, 9
923
NM_001323397.1
NP_001310326.1
8, 9
1091
NM_001323398.1
NP_001310327.1
7, 8
967
NM_001323399.1
NP_001310328.1
7, 8
834
NM_001323400.1
NP_001310329.1
6, 7
983
NM_001323401.1
NP_001310330.1
7, 8
1007
NM_001978.4
NP_001969.2
9, 10
1248
XM_005273432.1
XP_005273489.1
9, 10
980
XM_005273433.1
XP_005273490.1
9, 10
912
XM_005273442.1
XP_005273499.1
8, 9
764
XM_011544435.1
XP_011542737.1
8, 9
1173
XM_017013189.1
XP_016868678.1
9, 10
921
XM_017013190.1
XP_016868679.1
8, 9
1173
XM_017013191.1
XP_016868680.1
8, 9
927
XM_017013192.1
XP_016868681.1
8, 9
808
XM_017013193.1
XP_016868682.1
7, 8
940
XM_017013194.1
XP_016868683.1
7, 8
807
XM_017013195.1
XP_016868684.1
8, 9
1173
XM_017013196.1
XP_016868685.1
8, 9
776
XM_017013197.1
XP_016868686.1
8, 9
1007
XM_017013198.1
XP_016868687.1
8, 9
849
GenBank mRNA
AB209718.1
-
5, 6
474
AK055842.1
-
9, 10
912
AK091581.1
-
8, 9
927
AK289650.1
-
9, 10
880
AK295452.1
-
7, 8
853
BC006318.2
-
9, 10
997
4125804
BC017445.1
-
9, 10
997
3162732
BC052805.1
-
9, 10
1038
6269658
BT007396.1
-
8, 9
725
L19713.1
-
9, 10
1175
U28389.1
-
9, 10
1175
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108385, XM_008770831, XM_006252279, XM_006252281, XM_008770830, and others.
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
transmembrane transport
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
protein binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0