Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193476.1 | NP_001180405.1 | 17 | 2236 | ||
NM_052961.3 | NP_443193.1 | 17 | 2260 | |||
NM_138718.2 | NP_619732.2 | 15 | 1983 | |||
XM_011514294.2 | XP_011512596.1 | 16 | 2401 | |||
XM_017010235.1 | XP_016865724.1 | 17 | 2286 | |||
GenBank mRNA | AF314959.1 | - | 16 | 2130 | ||
AF331522.1 | - | 17 | 2204 | |||
AK055314.1 | - | 7 | 1706 | |||
AK057276.1 | - | 7 | 779 | |||
AK097410.1 | - | 11 | 1322 | |||
AK122928.1 | - | 17 | 2236 | |||
BC025408.2 | - | 15 | 1976 | 4823098 |