Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP1MT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258446.1 | NP_001245375.1 | 11 | 1399 | ||
NM_001258447.1 | NP_001245376.1 | 10 | 1258 | |||
NM_052963.2 | NP_443195.1 | 10 | 1307 | |||
XM_005250781.3 | XP_005250838.1 | 10 | 1295 | |||
GenBank mRNA | AF349017.1 | - | 10 | 1296 | ||
AK302372.1 | - | 10 | 1258 | |||
AK303237.1 | - | 9 | 1039 | |||
AK308475.1 | - | 10 | 1280 | |||
AK309005.1 | - | 10 | 1472 | |||
AK311647.1 | - | 7 | 773 | |||
AK315913.1 | - | 10 | 1269 | |||
AK316380.1 | - | 11 | 1346 | |||
BC044646.1 | - | 10 | 1282 | |||
BC052285.1 | - | 10 | 1282 | 5443410 | ||
BC071914.1 | - | 10 | 1280 | 6372930 |