Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014763.1 | NP_001014763.1 | 5 | 1235 | ||
NM_001985.2 | NP_001976.1 | 6 | 850 | |||
GenBank mRNA | AF370381.1 | - | 5 | 1217 | ||
AK055285.1 | - | 5 | 1237 | |||
AK128845.1 | - | 3535 | ||||
AK291881.1 | - | 6 | 777 | |||
AL833205.1 | - | 5 | 3511 | |||
BC093961.1 | - | 6 | 781 | 7939806 | ||
BC093963.1 | - | 6 | 780 | 7939808 | ||
BG281641.1 | - | 5 | 668 | |||
X71129.1 | - | 6 | 785 |