Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001241.3 | NP_001232.1 | 9 | 854 | ||
NM_001320748.1 | NP_001307677.1 | 10 | 948 | |||
NM_001320749.1 | NP_001307678.1 | 10 | 941 | |||
NM_058241.2 | NP_490595.1 | 9 | 854 | |||
NR_037649.1 | 10 | 941 | ||||
NR_135468.1 | 10 | 1260 | ||||
XM_017005227.1 | XP_016860716.1 | 8 | 803 | |||
XM_017005228.1 | XP_016860717.1 | 8 | 786 | |||
XM_017005229.1 | XP_016860718.1 | 10 | 1051 | |||
XM_017005230.1 | XP_016860719.1 | 10 | 1048 | |||
XM_017005231.1 | XP_016860720.1 | 10 | 1052 | |||
XM_017005232.1 | XP_016860721.1 | 10 | 1048 | |||
XM_017005233.1 | XP_016860722.1 | 5 | 3793 | |||
GenBank mRNA | AF048731.1 | - | 9 | 853 | ||
AF048732.1 | - | 9 | 821 | |||
AK292913.1 | - | 9 | 846 | |||
AK294889.1 | - | 10 | 941 | |||
BC114366.2 | - | 9 | 840 | 40053797 | ||
BX648174.1 | - | 10 | 1886 |