Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320747.1 | NP_001307676.1 | 22 | 3571 | ||
NM_001988.3 | NP_001979.2 | 22 | 3505 | |||
GenBank mRNA | BC126103.1 | - | 22 | 3284 | 8991819 | |
BC126105.1 | - | 22 | 3284 | 8991821 | ||
BC143814.1 | - | 22 | 3350 | 9052329 | ||
U53786.1 | - | 22 | 3349 |