Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080283.3 | NP_525022.2 | 11 | 1625 | ||
XM_017024011.1 | XP_016879500.1 | 11 | 1590 | |||
XM_017024012.1 | XP_016879501.1 | 11 | 1694 | |||
XM_017024013.1 | XP_016879502.1 | 11 | 1514 | |||
XM_017024014.1 | XP_016879503.1 | 11 | 2114 | |||
XM_017024015.1 | XP_016879504.1 | 8 | 1137 | |||
GenBank mRNA | AF423307.1 | - | 10 | 1482 | ||
AK057068.1 | - | 11 | 1557 | |||
AK074491.1 | - | 4 | 434 | |||
AY028899.1 | - | 11 | 1625 | |||
CR627382.1 | - | 10 | 1431 |