Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080283.3 | NP_525022.2 | 19 | 2649 | ||
XM_017024011.1 | XP_016879500.1 | 19 | 2614 | |||
XM_017024012.1 | XP_016879501.1 | 19 | 2718 | |||
XM_017024013.1 | XP_016879502.1 | 19 | 2538 | |||
XM_017024014.1 | XP_016879503.1 | 19 | 3138 | |||
XM_017024015.1 | XP_016879504.1 | 16 | 2161 | |||
GenBank mRNA | AF423307.1 | - | 18 | 2506 | ||
AK057068.1 | - | 19 | 2581 | |||
AK074491.1 | - | 12 | 1458 | |||
AY028899.1 | - | 19 | 2649 | |||
CR627382.1 | - | 18 | 2455 |