Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 20 | 3399 | ||
NM_001321081.1 | NP_001308010.1 | 21 | 3463 | |||
NM_001321082.1 | NP_001308011.1 | 20 | 3370 | |||
NM_001991.4 | NP_001982.2 | 21 | 3490 | |||
XM_005257145.2 | XP_005257202.1 | 19 | 3245 | |||
XM_011524517.2 | XP_011522819.1 | 20 | 3338 | |||
XM_017024350.1 | XP_016879839.1 | 20 | 3317 | |||
XM_017024351.1 | XP_016879840.1 | 19 | 3224 | |||
XM_017024352.1 | XP_016879841.1 | 18 | 3075 | |||
GenBank mRNA | AB002386.1 | - | 20 | 3402 | ||
AL833159.1 | - | 21 | 3410 | |||
BC015882.1 | - | 21 | 3425 | 3908776 |