Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289112.1 | NP_001276041.1 | 9 | 1098 | ||
NM_130808.2 | NP_570720.1 | 9 | 1726 | |||
NM_153429.1 | NP_702907.1 | 10 | 1086 | |||
XM_011512406.2 | XP_011510708.1 | 8 | 873 | |||
XM_011512407.2 | XP_011510709.1 | 8 | 1130 | |||
XM_011512408.2 | XP_011510710.1 | 8 | 2258 | |||
XM_017005693.1 | XP_016861182.1 | 12 | 2479 | |||
XM_017005694.1 | XP_016861183.1 | 8 | 850 | |||
XM_017005695.1 | XP_016861184.1 | 11 | 1349 | |||
XM_017005696.1 | XP_016861185.1 | 3 | 243 | |||
GenBank mRNA | AF465770.1 | - | 12 | 1426 | ||
AF465771.1 | - | 10 | 1086 | |||
AK056175.1 | - | 9 | 1198 | |||
AK128117.1 | - | 13 | 2976 | |||
AK295557.1 | - | 9 | 1013 | |||
BC014396.1 | - | 9 | 1284 | 3162811 | ||
BC075046.2 | - | 9 | 875 | 30915576 |