Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321917.1 | NP_001308846.1 | 5 | 2405 | ||
NM_133466.3 | NP_597723.1 | 5 | 2402 | |||
GenBank mRNA | AB075828.1 | - | 1 | 3869 | ||
AK074942.1 | - | 5 | 2384 | |||
AL834267.1 | - | 5 | 2376 | |||
BC032577.2 | - | 6 | 2342 | 5492906 | ||
GU388429.1 | - | 5 | 2372 |