Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF384 SIRNA Assays ›
Gene Symbol:
ZNF384
Gene Name:
zinc finger protein 384
Gene Aliases:
CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1
Chromosome Location:
Chr.12: 6666477 - 6689572 on Build GRCh38
UniGene ID:
Hs.103315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109974
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF384
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039920.2
NP_001035009.1
5
614
NM_001135734.2
NP_001129206.1
6
779
NM_133476.4
NP_597733.2
6
852
XM_011520917.1
XP_011519219.1
6
1070
XM_011520918.1
XP_011519220.1
6
779
XM_011520919.1
XP_011519221.1
6
820
XM_011520920.1
XP_011519222.1
6
804
XM_011520921.2
XP_011519223.1
6
629
XM_011520922.1
XP_011519224.1
5
731
XM_011520923.1
XP_011519225.1
5
756
XM_017018941.1
XP_016874430.1
6
804
XM_017018942.1
XP_016874431.1
6
670
XM_017018943.1
XP_016874432.1
6
664
XM_017018944.1
XP_016874433.1
5
731
XM_017018945.1
XP_016874434.1
5
756
XM_017018946.1
XP_016874435.1
6
804
XM_017018947.1
XP_016874436.1
6
820
XM_017018948.1
XP_016874437.1
4
1248
XM_017018949.1
XP_016874438.1
6
779
XM_017018950.1
XP_016874439.1
6
820
XM_017018951.1
XP_016874440.1
5
731
XM_017018952.1
XP_016874441.1
5
756
XM_017018953.1
XP_016874442.1
5
772
XM_017018954.1
XP_016874443.1
5
1387
GenBank mRNA
AK095734.1
-
6
801
AK298962.1
-
6
717
BC053361.1
-
5
496
6012540
BM554320.1
-
6
761
BX401142.2
-
5
725
DQ323638.1
-
1
66
DQ323639.1
-
1
66
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
nucleocytoplasmic transport
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
SH3 domain binding
sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0