Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144884.1 | NP_001138356.1 | 12 | 2572 | ||
NM_133496.4 | NP_598003.2 | 11 | 2886 | |||
XM_011540779.2 | XP_011539081.1 | 12 | 2985 | |||
GenBank mRNA | BC064692.1 | - | 11 | 2838 | 30330665 | |
BX537375.1 | - | 12 | 2568 | |||
BX648884.1 | - | 1 | 45 | |||
CR936806.1 | - | 12 | 2562 |