Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278734.1 | NP_001265663.1 | 8 | 2594 | ||
NM_016324.3 | NP_057408.2 | 5 | 2278 | |||
NM_016325.3 | NP_057409.1 | 7 | 2483 | |||
NM_133502.2 | NP_598009.1 | 8 | 2579 | |||
XM_011526327.1 | XP_011524629.1 | 7 | 2017 | |||
XM_011526328.2 | XP_011524630.1 | 3 | 1698 | |||
XM_017026174.1 | XP_016881663.1 | 6 | 2284 | |||
XM_017026175.1 | XP_016881664.1 | 6 | 2439 | |||
GenBank mRNA | AB029149.1 | - | 5 | 2238 | ||
AB029150.1 | - | 7 | 2385 | |||
AF275680.1 | - | 8 | 2258 | |||
AF275681.1 | - | 2 | 1692 | |||
AK095162.1 | - | 2 | 2923 | |||
AK222878.1 | - | 5 | 2206 | |||
BC009763.2 | - | 7 | 2106 | 2966764 | ||
BX641169.1 | - | 2 | 4693 | |||
CA942346.1 | - | 1 | 213 | |||
U71598.1 | - | 1 | 575 |