Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123226.1 | NP_001116698.1 | 10 | 1854 | ||
NM_012123.3 | NP_036255.2 | 9 | 1734 | |||
NM_133645.2 | NP_598400.1 | 10 | 1809 | |||
XM_006715444.2 | XP_006715507.1 | 9 | 1631 | |||
XM_006715445.2 | XP_006715508.1 | 9 | 1436 | |||
XM_006715446.3 | XP_006715509.1 | 9 | 1734 | |||
XM_011535723.1 | XP_011534025.1 | 10 | 1823 | |||
XM_011535724.1 | XP_011534026.1 | 10 | 1691 | |||
XM_011535725.2 | XP_011534027.1 | 10 | 1559 | |||
XM_017010697.1 | XP_016866186.1 | 9 | 1631 | |||
XM_017010698.1 | XP_016866187.1 | 10 | 1823 | |||
XM_017010699.1 | XP_016866188.1 | 8 | 1156 | |||
XM_017010700.1 | XP_016866189.1 | 8 | 1428 | |||
GenBank mRNA | AF132937.1 | - | 9 | 1656 | ||
AF319422.2 | - | 9 | 1700 | |||
AF469110.1 | - | 8 | 1409 | |||
AF469111.1 | - | 8 | 1790 | |||
AJ420530.1 | - | 8 | 1643 | |||
AK026073.1 | - | 5 | 718 | |||
AK074625.1 | - | 9 | 1713 | |||
AK225828.1 | - | 10 | 1838 | |||
AL833823.1 | - | 9 | 1578 | |||
AY078985.1 | - | 10 | 1775 | |||
AY078986.1 | - | 10 | 1775 | |||
BC005808.2 | - | 10 | 1494 | |||
BC011051.1 | - | 9 | 1675 | 4339769 |