Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282356.1 | NP_001269285.1 | 8, 9 | 1163 | ||
NM_052832.3 | NP_439897.1 | 7, 8 | 1124 | |||
NM_134266.1 | NP_599028.1 | 7, 8 | 1124 | |||
GenBank mRNA | AF331521.1 | - | 7, 8 | 1094 | ||
AJ413228.1 | - | 7, 8 | 1093 | |||
AJ413229.2 | - | 7, 8 | 1125 | |||
AJ413230.1 | - | 7, 8 | 1093 | |||
BC113866.1 | - | 6, 7 | 976 | 40034306 | ||
BC114474.1 | - | 6, 7 | 964 | 40034303 |