Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052934.3 | NP_443166.1 | 15 | 1762 | ||
NM_134325.2 | NP_599152.2 | 15 | 1762 | |||
XM_011509121.2 | XP_011507423.1 | 14 | 1625 | |||
XM_011509122.2 | XP_011507424.1 | 12 | 1314 | |||
GenBank mRNA | AF314958.1 | - | 14 | 1650 | ||
AF331525.1 | - | 15 | 1762 | |||
AK127491.1 | - | 7 | 1032 | |||
BC136538.1 | - | 15 | 1742 | 9020528 | ||
BC144233.1 | - | 14 | 1590 | |||
BC151208.1 | - | 15 | 1721 | 40147148 |