Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004019.1 | NP_001004019.1 | 18 | 4291 | ||
NM_001165035.1 | NP_001158507.1 | 18 | 4548 | |||
NM_001998.2 | NP_001989.2 | 17 | 4150 | |||
XM_006713026.2 | XP_006713089.1 | 16 | 4083 | |||
GenBank mRNA | AL050095.1 | - | 12 | 2201 | ||
AL080171.1 | - | 1 | 560 | |||
AY130459.1 | - | 18 | 4237 | |||
BC051690.1 | - | 17 | 4113 | 5269126 | ||
X82494.1 | - | 17 | 4096 |