Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052934.3 | NP_443166.1 | 21 | 4508 | ||
NM_134325.2 | NP_599152.2 | 22 | 4331 | |||
XM_011509121.2 | XP_011507423.1 | 20 | 4371 | |||
XM_011509122.2 | XP_011507424.1 | 18 | 4060 | |||
GenBank mRNA | AF314958.1 | - | 21 | 4219 | ||
AF331525.1 | - | 21 | 4508 | |||
AK091876.1 | - | 2 | 2154 | |||
AK127491.1 | - | 13 | 3778 | |||
BC151208.1 | - | 21 | 4467 | 40147148 |